Learn More
Alkali Scientific Toluidine Blue, 100 G
SDP
Catalog No. 5000661041
Encompass_Preferred
Toluidine Blue (100g) is a commonly used stain in histology, primarily for the examination of tissue sections and cell samples. It is effective in staining a wide variety of tissue types, including cartilage, mast cells, and other structures of interest in diagnostic histopathology. The 100g quantity is ideal for larger scale applications, offering consistent and high-quality results for laboratories with high throughput needs. Toluidine Blue produces distinct, vivid colors in tissue preparations, making it a valuable reagent for enhancing tissue visibility under the microscope. It is frequently used in both academic and clinical laboratories for routine histological examinations.
Catalog No. 50-006-61041 Supplier Alkali Scientific Supplier No. TOL004CP
Add to Cart
Add to Cart
Toluidine Blue (100g) is a commonly used stain in histology, primarily for the examination of tissue sections and cell samples. It is effective in staining a wide variety of tissue types, including cartilage, mast cells, and other structures of interest in diagnostic histopathology. The 100g quantity is ideal for larger scale applications, offering consistent and high-quality results for laboratories with high throughput needs. Toluidine Blue produces distinct, vivid colors in tissue preparations, making it a valuable reagent for enhancing tissue visibility under the microscope. It is frequently used in both academic and clinical laboratories for routine histological examinations.