Learn More
TARGETMOL CHEMICALS INC Toluidine Blue 100MG
SDP
Catalog No. 502880501
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Toluidine Blue (Blutene chloride), a phenothiazine, is used as a biological stain, a hemostatic, and a dye for silk and wool. Blutene chloride is also used as a diagnostic aid for gastric neoplasms and oral, and used in the identification of the parathyroid gland in thyroid surgery.
Catalog No. 50-288-0501 Supplier TARGETMOL CHEMICALS INC Supplier No. T0312100MG
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Toluidine Blue (Blutene chloride), a phenothiazine, is used as a biological stain, a hemostatic, and a dye for silk and wool. Blutene chloride is also used as a diagnostic aid for gastric neoplasms and oral, and used in the identification of the parathyroid gland in thyroid surgery.