Learn More
Alkali Scientific Toluidine Blue, 5 G
SDP
Catalog No. 5000661275
Encompass_Preferred
Toluidine Blue is a widely used histology stain available in a 5g format for tissue sectioning, microscopy, and cell analysis. This dye is particularly useful for staining cartilage, mast cells, and other tissue structures in histological studies. Toluidine Blue is known for its ability to produce clear, vivid contrasts in tissue samples, which aids in the identification and analysis of various biological structures under the microscope. It also plays a role in diagnostic pathology for detecting abnormal cell structures in tissue biopsies. This 5g package provides an ideal quantity for laboratories that need a high-quality, versatile stain for general histological applications.
Catalog No. 50-006-61275 Supplier Alkali Scientific Supplier No. TOL003CP
Toluidine Blue is a widely used histology stain available in a 5g format for tissue sectioning, microscopy, and cell analysis. This dye is particularly useful for staining cartilage, mast cells, and other tissue structures in histological studies. Toluidine Blue is known for its ability to produce clear, vivid contrasts in tissue samples, which aids in the identification and analysis of various biological structures under the microscope. It also plays a role in diagnostic pathology for detecting abnormal cell structures in tissue biopsies. This 5g package provides an ideal quantity for laboratories that need a high-quality, versatile stain for general histological applications.