Learn More
Medchemexpress LLC Toluidine blue | 92-31-9 | 250 MG
SDP
Catalog No. 5000414111
Encompass_Preferred
Toluidine Blue (Toluidine Blue O) is an alkaline quinonimine dye that has a high affinity for acidic tissue components, causing nuclei to stain blue and polysaccharides purple. It exhibits heterostaining properties for mast cells, mucins, and chondrocytes. It can also stain different components of plant tissues and cells in various colors. Toluidine Blue is also utilized as a diagnostic aid for identifying malignant lesions, such as cancer.
  • Fluorescent dye
  • Histological analysis
  • Histochemistry
  • Histochemistry dyes
  • Thiazine metachromatic dye
  • Stains acidic tissue
  • Used in living tissue
Catalog No. 50-004-14111 Supplier Medchemexpress LLC Supplier No. 250MGHYD0220
Toluidine Blue (Toluidine Blue O) is an alkaline quinonimine dye that has a high affinity for acidic tissue components, causing nuclei to stain blue and polysaccharides purple. It exhibits heterostaining properties for mast cells, mucins, and chondrocytes. It can also stain different components of plant tissues and cells in various colors. Toluidine Blue is also utilized as a diagnostic aid for identifying malignant lesions, such as cancer.
  • Fluorescent dye
  • Histological analysis
  • Histochemistry
  • Histochemistry dyes
  • Thiazine metachromatic dye
  • Stains acidic tissue
  • Used in living tissue