Learn More
Medchemexpress LLC Toluidine Blue mixture | 92-31-9 | 97.0% | 100 MG
SDP
Catalog No. 5000409762
Encompass_Preferred
Toluidine Blue is an alkaline quinonimine dye with high affinity for acidic tissue components, staining nuclei blue and polysaccharides purple. It exhibits heterostaining properties and can stain different components of plant tissues and cells in various colors. It is also used as a diagnostic aid for identifying malignant lesions like cancer.
  • Stains connective tissue mucins purple to red, with a blue background.
  • Stains mast cell granules purple.
  • Stains amyloid blue, showing bright red birefringence under polarized light.
  • Stains endocrine cell granules purple to red.
  • Stains sulfatides red brown or yellow.
  • Stains Corneybacterium diphtheria granules red violet.
  • Stains Helicobacter dark blue.
  • Used to stain frozen sections for rapid results and improved cell clarity.
  • Dyes plant lignins blue-green, phloem blue-purple, and other plant parts pale blue-green.
Catalog No. 50-004-09762 Supplier Medchemexpress LLC Supplier No. HYD0220A100MG
Add to Cart
Add to Cart
Toluidine Blue is an alkaline quinonimine dye with high affinity for acidic tissue components, staining nuclei blue and polysaccharides purple. It exhibits heterostaining properties and can stain different components of plant tissues and cells in various colors. It is also used as a diagnostic aid for identifying malignant lesions like cancer.
  • Stains connective tissue mucins purple to red, with a blue background.
  • Stains mast cell granules purple.
  • Stains amyloid blue, showing bright red birefringence under polarized light.
  • Stains endocrine cell granules purple to red.
  • Stains sulfatides red brown or yellow.
  • Stains Corneybacterium diphtheria granules red violet.
  • Stains Helicobacter dark blue.
  • Used to stain frozen sections for rapid results and improved cell clarity.
  • Dyes plant lignins blue-green, phloem blue-purple, and other plant parts pale blue-green.