Learn More
Description
- Effectively visualize cartilaginous and chondrogenic differentiated tissues with the blue-colored stain solution: Toluidine Blue O, 1% aqueous.
Specifications
Specifications
| Melting Point | ~0°C (32°F in water) |
| Boiling Point | ~100°C (212°F in water) |
| CAS Min % | 1%, 99% |
| Concentration | 1% |
| Dye Content | 1% |
| Extinction Coefficient | 13000 M-1 cm-1 |
| Identification | Toluidine Blue O, 1% Aqueous |
| CAS Max % | 99%, 1% |
| Infrared Spectrum | Yes |
| Lambda Max | 241-247 nm |
| Show More |
Safety and Handling
| Hazard Category | May be harmful if swallowed. |
| Hazard Statement | May cause respiratory irritation. |
| Precautionary Statement | May cause eye irritation. |
| ShelfLife | 3 years |
| DOTInformation | Not regulated. |
| EINECSNumber | 202-146-2 |
| RTECSNumber | SP5670000 |
| Recommended Storage | Store in a cool, well-ventilated area away from incompatible substances. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
