Learn More
Alkali Scientific Toluidine Blue O, 1 Kg
SDP
Catalog No. 5000660888
Encompass_Preferred
Toluidine Blue O, 1kg is a powerful histology staining dye used to visualize cell structures in tissue samples. The dye has a particular affinity for acidic tissue components, such as mucopolysaccharides and nucleic acids, making it particularly useful in tissue diagnostics and pathology. It is commonly employed in histological staining procedures to highlight tissue morphology and cell viability. The 1kg pack size is ideal for research laboratories and institutions conducting large-scale or frequent histology studies. This dye is particularly useful for visualizing mast cells, detecting tumors, and assessing tissue composition in a variety of biological and medical research applications.
Catalog No. 50-006-60888 Supplier Alkali Scientific Supplier No. TOL001CP
Add to Cart
Add to Cart
Toluidine Blue O, 1kg is a powerful histology staining dye used to visualize cell structures in tissue samples. The dye has a particular affinity for acidic tissue components, such as mucopolysaccharides and nucleic acids, making it particularly useful in tissue diagnostics and pathology. It is commonly employed in histological staining procedures to highlight tissue morphology and cell viability. The 1kg pack size is ideal for research laboratories and institutions conducting large-scale or frequent histology studies. This dye is particularly useful for visualizing mast cells, detecting tumors, and assessing tissue composition in a variety of biological and medical research applications.