Learn More
Bioworld Toluidine Blue O, 25 g
Catalog No. 50199239
Encompass_Preferred
Higth Purity Grade A metachromatic stain for histology. EM (626nm, water) >45,000 Moisture <5.0%
Catalog No. 50-199-239 Supplier Bioworld Supplier No. 217700321
Higth Purity Grade A metachromatic stain for histology. EM (626nm, water) >45,000 Moisture <5.0%