Learn More
Alkali Scientific Toluidine Blue O, 25 G
SDP
Catalog No. 5000660889
Encompass_Preferred
Toluidine Blue O, 25g is a histological stain used to identify acidic tissue components like glycosaminoglycans, mucopolysaccharides, and nucleic acids in tissue samples. The dye’s affinity for acidic structures allows it to selectively stain mast cells, cartilage, and certain connective tissues, making it an essential tool for pathologists. This dye is commonly applied in the preparation of tissue slides for microscopy in histological studies, particularly in examining connective tissue and tumors. The 25g pack size provides ample dye for routine lab use, making it an excellent choice for laboratories and medical facilities requiring efficient and cost-effective staining for tissue analysis.
Catalog No. 50-006-60889 Supplier Alkali Scientific Supplier No. TOL002CP
Toluidine Blue O, 25g is a histological stain used to identify acidic tissue components like glycosaminoglycans, mucopolysaccharides, and nucleic acids in tissue samples. The dye’s affinity for acidic structures allows it to selectively stain mast cells, cartilage, and certain connective tissues, making it an essential tool for pathologists. This dye is commonly applied in the preparation of tissue slides for microscopy in histological studies, particularly in examining connective tissue and tumors. The 25g pack size provides ample dye for routine lab use, making it an excellent choice for laboratories and medical facilities requiring efficient and cost-effective staining for tissue analysis.