Learn More
Description
Certified for use as a metachromatic stain for mast cells and cartilage using the Churukian-Schenk method and the method of Nocht on paraffin sections.
Specifications
Specifications
| Chemical Name or Material | Toluidine Blue O |
| CAS | 92-31-9 |
| Molecular Weight (g/mol) | 305.83 g/mol |
| Grade | Certified |
| Color | Dark Green |
| Quantity | 5 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.