Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Toluidine Blue O, pure

Catalog No. AC348601000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348601000 100 g Glass bottle
AC348600050 5 g Glass bottle
AC348600250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC348601000 Supplier Thermo Scientific Chemicals Supplier No. 348601000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-31-9
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
Synonym Basic blue 17, C.I. 52040, Tolonium chloride
PubChem CID 7083
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Beilstein 27, 402
Merck Index 15, 9679
Solubility Information Solubility in water: 38.2g/L. Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
Formula Weight 305.82
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Toluidine blue O
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.