Learn More
Crescent Chemical Co Inc TOLUIDINE BLUE O SALT
SDP
Catalog No. 50248202
Encompass_Preferred
Toluidine Blue O salt Methylhomologue of Azure A. For RNA staining and RNase detection in electrophoresis (1). Stain for oligodeoxyribonucleotides (2). Stain for acidic mucopolysaccharides (proteoglycans) (3) and RNA (4). Size - 25G Storage Conditions - +15 °
Catalog No. 50-248-202 Supplier Crescent Chemical Co Inc Supplier No. SE36693.02
Toluidine Blue O salt Methylhomologue of Azure A. For RNA staining and RNase detection in electrophoresis (1). Stain for oligodeoxyribonucleotides (2). Stain for acidic mucopolysaccharides (proteoglycans) (3) and RNA (4). Size - 25G Storage Conditions - +15 °