Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Toluidine Blue O Solution, 0.5%
SCI_ED

Catalog No. S25613 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
15 mL
100 mL
Packaging:
0.5%
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25613 100 mL 0.5%
S25612 15 mL 0.5%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25613

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 92-31-9 , 7732-18-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
PubChem CID 7083
IUPAC Name 3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Assay Percent Range 0.5% w/v
Grade Laboratory
Packaging 0.5%
Density ∽1g/mL
Color Blue/Purple
Quantity 100 mL
Chemical Name or Material Toluidine Blue O Solution, 0.5%
Safety handling Check Alert storage code Red
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.