Description
Toluidine Blue O (TBO) is a thiazine dye of the quinone-imine family and is cationic in nature. It has been employed in polychromatic staining of paraffin embedded plant cell walls. It is a metachromatic dye suitable for a wide variety of histological staining procedures.
Specifications
Specifications
| CAS | 92-31-9 |
| Quantity | 100 g |
| Product Type | Toluidine Blue |