Learn More
Toluidine Blue O, Technical Grade, Sigma-Aldrich
Catalog No. 01259071 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-071 5 g
01-259-068 100 g
01-259-070 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-259-071 Supplier MilliporeSigma Supelco Supplier No. T32605G
Add to Cart
Add to Cart
Metachromatic thiazine dye suitable for polychromatic and histological staining.
Toluidine Blue O (TBO) is a thiazine dye of the quinone-imine family and is cationic in nature. It has been employed in polychromatic staining of paraffin embedded plant cell walls. It is a metachromatic dye suitable for a wide variety of histological staining procedures.
TRUSTED_SUSTAINABILITY
Specifications
CAS 92-31-9
Quantity 5 g
Product Type Toluidine Blue