Learn More
Toluidine Blue Stain, 1% (w/v) in 20% (v/v) Alcohol with 1% (v/v) Acetic Acid, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | TOLUIDINE BLUE |
| CAS | 7732-18-5, 64-17-5, 64-19-7, 92-31-9, 67-56-1 |
| CAS Min % | 83.0, 13.28, 0.87, 0.83 |
| CAS Max % | 86.0, 13.82, 0.91, 0.86 |
| Packaging | Amber Glass Bottle |
| Color | Dark Blue |
| pH | 2.8 |
| Quantity | 500 mL |
| Percent Purity | 1% W/V |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.