Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluidine Blue Stain, 1% (w/v), Ricca Chemical
SDP

Catalog No. p-3252591 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
865416 500 mL Amber Glass Bottle
865432 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 865416 Supplier Ricca Chemical Company Supplier No. 865416
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-31-9 , 7732-18-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
PubChem CID 7083
IUPAC Name 3-amino-7-(dimethylamino)-2-methyl-5λ4-phenothiazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Color Blue/Purple
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Toluidine Blue Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.