Learn More
Medchemexpress LLC Toluidine blue (ZnCl2) | 6586-04-5 | 84.5% | 654.6 g/mol | C15H16N3S·xCl2Zn·Cl | 100 MG
SDP
Catalog No. 5000282073
Encompass_Preferred
Toluidine blue (ZnCl2) is a basic thiazine dye supplied as the zinc chloride double salt, commonly used as a biological stain for microscopy and biochemical assays. It preferentially stains acidic tissue components such as nucleic acids, making it useful for histology, cytology, and diagnostic staining.
  • Basic thiazine dye for staining nucleic acids and acidic tissue components.
  • Supplied as the zinc chloride double salt suitable for staining protocols.
  • Commonly used in microscopy, histology, and cytology applications.
  • Assay purity reported as 84.5% (manufacturer data).
  • Supplied in laboratory-scale quantities for research use.
Catalog No. 50-002-82073 Supplier Medchemexpress LLC Supplier No. HYW250143100MG
Toluidine blue (ZnCl2) is a basic thiazine dye supplied as the zinc chloride double salt, commonly used as a biological stain for microscopy and biochemical assays. It preferentially stains acidic tissue components such as nucleic acids, making it useful for histology, cytology, and diagnostic staining.
  • Basic thiazine dye for staining nucleic acids and acidic tissue components.
  • Supplied as the zinc chloride double salt suitable for staining protocols.
  • Commonly used in microscopy, histology, and cytology applications.
  • Assay purity reported as 84.5% (manufacturer data).
  • Supplied in laboratory-scale quantities for research use.