Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Toluidine blue (ZnCl2) | 6586-04-5 | MFCD03548293 | 84.5% | 373.97 g/mol | C15H16ClN3S · 0.5ZnCl2 | 1 G
SDP

Catalog No. 5000280864
Encompass_Preferred

Toluidine blue (ZnCl2) is a basic thiazine dye supplied as the zinc chloride salt for histology and microscopy staining. It preferentially stains nucleic acids, mast cells, and other acidic tissue components; supplied as a solid powder with defined assay and storage recommendations for laboratory research use.

  • Basic thiazine dye for nucleic acid and mast cell staining.
  • Zinc chloride salt form for consistent staining properties.
  • Assay 84.5% provides defined staining performance.
  • Brown to black solid powder suitable for solution preparation.
  • Supplied in 1 g packages for laboratory use.
  • Store at -20°C and protect from light to preserve stability.
  • SDS available for handling, storage, and safety information.

Catalog No. 50-002-80864 Supplier Medchemexpress LLC Supplier No. HYW2501431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toluidine blue (ZnCl2) is a basic thiazine dye supplied as the zinc chloride salt for histology and microscopy staining. It preferentially stains nucleic acids, mast cells, and other acidic tissue components; supplied as a solid powder with defined assay and storage recommendations for laboratory research use.

  • Basic thiazine dye for nucleic acid and mast cell staining.
  • Zinc chloride salt form for consistent staining properties.
  • Assay 84.5% provides defined staining performance.
  • Brown to black solid powder suitable for solution preparation.
  • Supplied in 1 g packages for laboratory use.
  • Store at -20°C and protect from light to preserve stability.
  • SDS available for handling, storage, and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.