Learn More
Medchemexpress LLC Toluidine blue (ZnCl2) | 6586-04-5 | MFCD03548293 | 84.5% | 373.97 g/mol | C15H16ClN3S·0.5ZnCl2 | 50 MG
SDP
Catalog No. 5000280869
Encompass_Preferred
Toluidine blue (ZnCl2) is a basic thiazine dye used for biological staining in microscopy and histology. It has high affinity for acidic tissue components and produces a deep bluish-purple metachromatic stain, making it suitable for general staining, diagnostic screening, and connective tissue visualization.
  • High affinity for acidic tissue components such as nucleic acids.
  • Metachromatic staining highlights mast cells and connective tissue.
  • Provided as a ZnCl2 complex for enhanced stability.
  • Suitable for microscopy and histological applications.
  • Available in small research quantities (for example, 50 mg).
  • Assay purity reported as 84.5% by supplier.
Catalog No. 50-002-80869 Supplier Medchemexpress LLC Supplier No. HYW25014350MG
Toluidine blue (ZnCl2) is a basic thiazine dye used for biological staining in microscopy and histology. It has high affinity for acidic tissue components and produces a deep bluish-purple metachromatic stain, making it suitable for general staining, diagnostic screening, and connective tissue visualization.
  • High affinity for acidic tissue components such as nucleic acids.
  • Metachromatic staining highlights mast cells and connective tissue.
  • Provided as a ZnCl2 complex for enhanced stability.
  • Suitable for microscopy and histological applications.
  • Available in small research quantities (for example, 50 mg).
  • Assay purity reported as 84.5% by supplier.