Learn More
Selleck Chemical LLC Tolvaptan-10mg
SDP
Supplier:  Selleck Chemical LLC S259310MG
Encompass_Preferred
Tolvaptan (OPC-41061) is an orally effective nonpeptide arginine vasopressin V2 receptor antagonist with IC50 of 3 nM, used to treat hyponatremia.
Catalog No. 50-239-0079