Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tolylene-2,4-diisocyanate

Catalog No. 2168361KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-683-61KG 1 kg
21-683-6250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-683-61KG Supplier Sigma Aldrich Supplier No. 2168361KG
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tolylene-2,4-diisocyanate

Specifications

CAS 584-84-9
Melting Point 20°C to 22°C (lit.)
Density 1.225 g/mL (at 25°C)
Boiling Point 115°C to 120°C (10 mmHg)
Molecular Formula C9H6N2O2
Refractive Index n20/D 1.568 (literature)
Linear Formula CH3C6H3(NCO)2
MDL Number MFCD00002011
Quantity 1 kg
Synonym 2,4-Diisocyanatotoluene; 4-Methyl-1,3-phenylene diisocyanate; 4-Methyl-m-phenylene diisocyanate; BASF LUPRANATE T80; TDI
Molecular Weight (g/mol) 174.16
Percent Purity 80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.