Learn More
Kuhner Shaker Inc TOM ADD-ON KIT - PUMP VERSION.
Catalog No. 502570065
Encompass
TOM Add-on Kit - Pump Version. Measures OTR, CTR and RQ in 4 shake flasks; Contains: TOM Sensor module - pump aeration 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2, 0-25% O2; connection cable 0.5m; 4 250mL TOM shake flasks included
Catalog No. 50-257-0065 Supplier Kuhner Shaker Inc Supplier No. SMTO50007
Add to Cart
Add to Cart
Request a Quote
TOM Add-on Kit - Pump Version. Measures OTR, CTR and RQ in 4 shake flasks; Contains: TOM Sensor module - pump aeration 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2, 0-25% O2; connection cable 0.5m; 4 250mL TOM shake flasks included