Learn More
Kuhner Shaker Inc TOM MASSFLOW CONTROLLER ADD-ON
Catalog No. 502570075
Encompass
TOM Massflow controller add-on:..Massflow controller 0-1nL/min with accessories for TOM Supply module, only works with TOM supply module; max. pressure 2bar....TOM Supply Modul is necessary..
Catalog No. 50-257-0075 Supplier Kuhner Shaker Inc Supplier No. SMTO30002
Add to Cart
Add to Cart
Request a Quote
TOM Massflow controller add-on:..Massflow controller 0-1nL/min with accessories for TOM Supply module, only works with TOM supply module; max. pressure 2bar....TOM Supply Modul is necessary..