Learn More
Kuhner Shaker Inc TOM MEASUREMENT MODULE- MFC ..
Catalog No. 502570073
Encompass
TOM measurement module- MFC:....Measurement module to measure OTR, CTR and RQ in 4 shake flasks; 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2, 0-25% O2;..
Catalog No. 50-257-0073 Supplier Kuhner Shaker Inc Supplier No. SMTO30004
Add to Cart
Add to Cart
Request a Quote
TOM measurement module- MFC:....Measurement module to measure OTR, CTR and RQ in 4 shake flasks; 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2, 0-25% O2;..