Learn More
Kuhner Shaker Inc TOM MEASUREMENT MODULE- PUMP..
Catalog No. 502570074
Encompass
TOM Measurement Module- Pump....Measurement module to measure OTR, CTR and RQ in 4 shake flasks; 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2,..0-25% O2;....Needs: TOM supply module, TOM mount..
Catalog No. 50-257-0074 Supplier Kuhner Shaker Inc Supplier No. SMTO30003
Add to Cart
Add to Cart
Request a Quote
TOM Measurement Module- Pump....Measurement module to measure OTR, CTR and RQ in 4 shake flasks; 377x99x102(LxBxH), ambient pressure, 15-40°C, 30-85% rel. humidity, 0-16% CO2,..0-25% O2;....Needs: TOM supply module, TOM mount..