Learn More
Kuhner Shaker Inc TOM STARTER KIT - MFC VERSION.
Catalog No. 502570066
Encompass
TOM Starter Kit - MFC Version. Measure OTR, CTR and RQ in 4 shake flasks; Contains: TOM supply module 342x136x93 (LxBxH), 100-260V, input voltage, USB 2.0 with TOM massflow controller add-on, max pressure 2bar;TOM measurement module (1x).
Catalog No. 50-257-0066 Supplier Kuhner Shaker Inc Supplier No. SMTO50004
Add to Cart
Add to Cart
Request a Quote
TOM Starter Kit - MFC Version. Measure OTR, CTR and RQ in 4 shake flasks; Contains: TOM supply module 342x136x93 (LxBxH), 100-260V, input voltage, USB 2.0 with TOM massflow controller add-on, max pressure 2bar;TOM measurement module (1x).