Learn More
Kuhner Shaker Inc TOM SUPPLY MODULE MODULE CONT
Catalog No. 502570072
Encompass
TOM Supply Module: Module contains power supply and USB to CAN interface to run up to 4 TOM measurement modules with TOM control software; 342x136x93 (LxBxH),100-260V input voltage, USB 2.0 (1x per machine)..
Catalog No. 50-257-0072 Supplier Kuhner Shaker Inc Supplier No. SMTO30011
Add to Cart
Add to Cart
Request a Quote
TOM Supply Module: Module contains power supply and USB to CAN interface to run up to 4 TOM measurement modules with TOM control software; 342x136x93 (LxBxH),100-260V input voltage, USB 2.0 (1x per machine)..