Learn More
Aviva Systems Biology Corp TOM1 Antibody - middle region (ARP52077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TOM1. Alternate names: FLJ33404ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53857