Learn More
Aviva Systems Biology Corp TOM1L1 Antibody - C-terminal region (ARP61882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOM1L1. It was validated on Western Blot. . Alternate names: OK/KNS-CL.3, SRCASMApplicationWB. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-287-497