Learn More
ABclonal Technology TOM20 Rabbit mAb 100 L
SDP
Supplier:  ABclonal Technology A19403
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 20-145 of human TOM20 (NP_055580.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC,IF,IP•Swiss Prot: Q15388•Gene ID:9804•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.09% sodium azide,0.05% BSA,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. NC2079874