Learn More
ABclonal Technology TOM70 Rabbit mAb
SDP
Catalog No. 5000005725
Encompass_Preferred
This gene encodes an import receptor of the outer mitochondrial membrane that is part of the translocase of the outer membrane complex This protein is involved in the import of mitochondrial precursor proteins The protein interacts with the SARS-CoV and SARS-Cov-2 ORF9b proteins
Catalog No. 50-000-05725 Supplier ABclonal Technology Supplier No. A2121020UL
This gene encodes an import receptor of the outer mitochondrial membrane that is part of the translocase of the outer membrane complex This protein is involved in the import of mitochondrial precursor proteins The protein interacts with the SARS-CoV and SARS-Cov-2 ORF9b proteins