Learn More
Medchemexpress LLC Tomatidine hydrochloride | 6192-62-7 | 50 MG
SDP
Catalog No. 5000421622
Encompass_Preferred
Tomatidine hydrochloride acts as an anti-inflammatory agent by blocking NF-κB and JNK signaling. It also activates autophagy in mammal cells or C elegans. In vitro, Tomatidine decreases inducible NO synthase and COX-2 expression by suppressing I-κBα phosphorylation, NF-κB nuclear translocation, and JNK activation, which subsequently inhibits c-jun phosphorylation and Oct-2 expression. It causes dose-dependent inhibition of LPS-induced iNOS expression and suppresses the binding activity of NF-κB.
  • Acts as an anti-inflammatory agent
  • Blocks NF-κB and JNK signaling pathways
  • Activates autophagy in mammal cells or C elegans
  • Decreases inducible NO synthase and COX-2 expression
  • Inhibits I-κBα phosphorylation and NF-κB nuclear translocation
  • Suppresses JNK activation and c-jun phosphorylation
  • Inhibits LPS-induced iNOS expression
  • Suppresses NF-κB binding activity
Catalog No. 50-004-21622 Supplier Medchemexpress LLC Supplier No. HYN2149A50MG
Tomatidine hydrochloride acts as an anti-inflammatory agent by blocking NF-κB and JNK signaling. It also activates autophagy in mammal cells or C elegans. In vitro, Tomatidine decreases inducible NO synthase and COX-2 expression by suppressing I-κBα phosphorylation, NF-κB nuclear translocation, and JNK activation, which subsequently inhibits c-jun phosphorylation and Oct-2 expression. It causes dose-dependent inhibition of LPS-induced iNOS expression and suppresses the binding activity of NF-κB.
  • Acts as an anti-inflammatory agent
  • Blocks NF-κB and JNK signaling pathways
  • Activates autophagy in mammal cells or C elegans
  • Decreases inducible NO synthase and COX-2 expression
  • Inhibits I-κBα phosphorylation and NF-κB nuclear translocation
  • Suppresses JNK activation and c-jun phosphorylation
  • Inhibits LPS-induced iNOS expression
  • Suppresses NF-κB binding activity