Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tomatidine hydrochloride | 6192-62-7 | MFCD00133866 | 99.6% | 452.11 | C27H46ClNO2 | 10 MG
SDP

Catalog No. 5000296040
Encompass_Preferred

Tomatidine hydrochloride is the hydrochloride salt of tomatidine, a steroidal alkaloid used as a research reagent that inhibits NF-κB and modulates JNK signaling and has been reported to induce autophagy in mammalian cells and Caenorhabditis elegans. It is supplied as a high-purity small molecule for laboratory use.

  • Hydrochloride salt, molecular formula C27H46ClNO2.
  • Molecular weight 452.11.
  • CAS number 6192-62-7.
  • Purity 99.6% (reported).
  • Reported activities: NF-κB inhibition, JNK modulation, autophagy induction.
  • Suitable for cell biology and signaling pathway studies.
  • Store sealed at 4°C, protected from moisture and light.

Catalog No. 50-002-96040 Supplier Medchemexpress LLC Supplier No. HYN2149A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tomatidine hydrochloride is the hydrochloride salt of tomatidine, a steroidal alkaloid used as a research reagent that inhibits NF-κB and modulates JNK signaling and has been reported to induce autophagy in mammalian cells and Caenorhabditis elegans. It is supplied as a high-purity small molecule for laboratory use.

  • Hydrochloride salt, molecular formula C27H46ClNO2.
  • Molecular weight 452.11.
  • CAS number 6192-62-7.
  • Purity 99.6% (reported).
  • Reported activities: NF-κB inhibition, JNK modulation, autophagy induction.
  • Suitable for cell biology and signaling pathway studies.
  • Store sealed at 4°C, protected from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.