Learn More
Medchemexpress LLC Tomatine | 17406-45-0 | 99.9% | 1034.19 g·mol⁻¹ | C50H83NO21 | 10 MG
SDP
Catalog No. 5000295602
Encompass_Preferred
Tomatine is a naturally occurring glycoalkaloid derived from tomato plants used as a biochemical research reagent. It has demonstrated bioactivity including induction of apoptosis via upregulation of apoptosis-inducing factor (AIF) and inhibition of 20S proteasome activity, and is supplied as a solid with defined storage recommendations for powder and solvent forms.
  • Natural glycoalkaloid derived from tomatoes.
  • Listed purity 99.90%.
  • Available in small research sizes including 10 mg and solution formats.
  • Shows apoptotic activity and proteasome inhibition in cell studies.
  • Powder and solvent storage conditions provided for stability.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-95602 Supplier Medchemexpress LLC Supplier No. HYN216610MG
Tomatine is a naturally occurring glycoalkaloid derived from tomato plants used as a biochemical research reagent. It has demonstrated bioactivity including induction of apoptosis via upregulation of apoptosis-inducing factor (AIF) and inhibition of 20S proteasome activity, and is supplied as a solid with defined storage recommendations for powder and solvent forms.
  • Natural glycoalkaloid derived from tomatoes.
  • Listed purity 99.90%.
  • Available in small research sizes including 10 mg and solution formats.
  • Shows apoptotic activity and proteasome inhibition in cell studies.
  • Powder and solvent storage conditions provided for stability.
  • For research use only; not for human therapeutic use.