Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tomatine from Tomato 80.0+%, TCI America™
SDP

Catalog No. T3636100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3636100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3636100MG Supplier TCI America Supplier No. T3636100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17406-45-0
Molecular Formula C50H83NO21
Molecular Weight (g/mol) 1034.2
MDL Number MFCD00016888
InChI Key REJLGAUYTKNVJM-MERLFABPSA-N
Synonym tomatine, 22r,25r-5alpha-spirosolane-3beta-yl 4-o-2-o-beta-d-glucopyranosyl-3-o-beta-d-xylopyranosyl-beta-d-glucopyranosyl-beta-d-galactopyranoside
PubChem CID 133612499
SMILES CC1CCC2(C(C3C(O2)CC4C3(CCC5C4CCC6C5(CCC(C6)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)O)OC9C(C(C(CO9)O)O)O)OC2C(C(C(C(O2)CO)O)O)O)O)O)C)C)C)NC1

Specifications

Melting Point 291°C
Quantity 100 mg
Formula Weight 1034.20
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tomatine from Tomato
RTECSNumber XW1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.