Learn More
NEOGEN Tomato Juice Agar, 500 g
SDP
Catalog No. 502015930
Encompass_Preferred
Tomato Juice Agar is used for the cultivation of lactobacilli in a laboratory setting. Tomato Juice Agar is not intended for use in the diagnosis of disease or other conditions in humans. In 1925, Mickle and Breed reported the use of tomato juice in culture media used for cultivating lactobacilli. Kulp investigated tomato juice on bacterial development and found growth of L. acidophilus was enhanced. Tomato Juice Agar, prepared according to Kulp and White’s modification, is especially useful in cultivating L. acidophilus from foodstuffs.For more information visit https://www.neogen.com/solutions/Microbiology/tomato-juice-agar
Catalog No. 50-201-5930 Supplier NEOGEN Supplier No. 700003648
Tomato Juice Agar is used for the cultivation of lactobacilli in a laboratory setting. Tomato Juice Agar is not intended for use in the diagnosis of disease or other conditions in humans. In 1925, Mickle and Breed reported the use of tomato juice in culture media used for cultivating lactobacilli. Kulp investigated tomato juice on bacterial development and found growth of L. acidophilus was enhanced. Tomato Juice Agar, prepared according to Kulp and White’s modification, is especially useful in cultivating L. acidophilus from foodstuffs.For more information visit https://www.neogen.com/solutions/Microbiology/tomato-juice-agar