Learn More
MOLTOX Molecular Toxicology Inc Tomato Juice Agar, 20 mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948699
Encompass_Preferred
Tomato Juice Agar is used for the cultivation and enumeration of the Lactobacillus species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-699 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S90.141
Tomato Juice Agar is used for the cultivation and enumeration of the Lactobacillus species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.