Learn More
MOLTOX Molecular Toxicology Inc Tomato Juice Broth, 100 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948770
Encompass_Preferred
Tomato Juice Broth us used for cultivating aciduric microorgansisms such as yeasts. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-770 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26584
Tomato Juice Broth us used for cultivating aciduric microorgansisms such as yeasts. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.