Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEOGEN TOMATO TASTE TESTING BROTH 5 K
SDP

Catalog No. NC1465154
Encompass_Preferred

TOMATO TASTE TESTING BROTH 5kg

Catalog No. NC1465154 Supplier NEOGEN Supplier No. 700002460
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TOMATO TASTE TESTING BROTH 5kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.