Learn More
Med Vet International Tomcat Catheter open end 3.5 Fr x 140 mm, J0153P, Each
Catalog No. 5000583715
Encompass_Preferred
JorVet Cat Catheter with Open End Hole • The catheter with open end hole is useful for loosening an obstruction by the pressure of the injection • Extra rigid for easy introduction Cat Catheter with open end hole, without stylet, 3.5fr. x 140mm
Catalog No. 50-005-83715 Supplier Med Vet International Supplier No. JOR153P
JorVet Cat Catheter with Open End Hole • The catheter with open end hole is useful for loosening an obstruction by the pressure of the injection • Extra rigid for easy introduction Cat Catheter with open end hole, without stylet, 3.5fr. x 140mm