Learn More
Medchemexpress LLC MedChemExpress | Tominersen | 5 mg | HY-132579-5mg | Purity: 97.34% | 1709886-74-7 | MW: 7078.00
SDP
Supplier:  Medchemexpress LLC HY1325795MG
Encompass_Preferred
Tominersen (RG6042) is a second-generation 2′-O-(2-methoxyethyl) antisense oligonucleotide that targets huntingtin protein (HTT) mRNA and potently suppresses HTT production. Tominersen improves survival and reduces brain atrophy in mice. Tominersen can be used for the research of Huntington’s disease (HD)
Catalog No. NC2743944