Learn More
Biolegend TotalSeq™-B0238 Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400581
Encompass_Preferred
TotalSeq™-B0238 Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: PG; Size: 10 ug
Catalog No. 50-207-9525