Learn More
Biolegend TotalSeq™-C0584 anti-human TCR Vα24-Jα18 (iNKT cell)
Supplier:  Biolegend 342925
Encompass_Preferred
TotalSeq™-C0584 anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactivity: Pigtailed Macaque, Sooty Mangabey; Apps: PG; Size: 10 ug
Catalog No. 50-207-6544