Learn More
ABclonal Technology TOMM22 Rabbit pAb
SDP
Catalog No. 5000106910
Encompass_Preferred
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.
Catalog No. 50-001-06910 Supplier ABclonal Technology Supplier No. A14548500UL
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.