Learn More
Aviva Systems Biology Corp TOMM34 Antibody - N-terminal region (ARP52257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOMM34. It was validated on Western Blot. . Alternate names: HTOM34P, TOM34, URCC3ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-675