Learn More
Aviva Systems Biology Corp TOMM40L Antibody - middle region (ARP35373_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOMM40L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TOMM40L. Alternate names: FLJ12770, RP11-297K8.10, TOMM40BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55716