Learn More
Electron Microscopy Sciences Adhesion Slide, Color: White. Corner: 45°., 100/PK
SDP
Catalog No. 502090848
Encompass_Preferred
These TOMO hydrophilic adhesion slides offer superior tissue adhesion, staining performance and slide surface fluidics. Proprietary slide coating ensures consistent performance even after exposure to heat stress and humid environmental conditions. Optimized for use with thermal printers. These slides are recommended for use on Ventana Benchmark staining platform and automated IHC platforms. They are exceptionally flat, with low fluorescence glass, and finished with 90 degree and clipped corners. Color: White. Corner: 45° .
Catalog No. 50-209-0848 Supplier Electron Microscopy Sciences Supplier No. 6370503
These TOMO hydrophilic adhesion slides offer superior tissue adhesion, staining performance and slide surface fluidics. Proprietary slide coating ensures consistent performance even after exposure to heat stress and humid environmental conditions. Optimized for use with thermal printers. These slides are recommended for use on Ventana Benchmark staining platform and automated IHC platforms. They are exceptionally flat, with low fluorescence glass, and finished with 90 degree and clipped corners. Color: White. Corner: 45° .