Learn More
Newcomer Supply TOMO Adhesive Microscope Slides, White - 90° Corners (1,000 slides/case)
SDP
Catalog No. NC2101966
Encompass
The advanced technology in manufacturing the adhesive surface of the TOMO microscope slide produces outstanding adhesion to the tissue section even in the most demanding applications. The surface of the TOMO adhesive microscope slide is hydrophilic producing two very notable features: allows to refloat the tissue in the water bath for repositioning the tissue section and allows the microscope slide to be used on all automated IHC platforms. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 90° Corner; Color: White; Unit: 100 slides/box, 1,000 slides/case. ISO 13485 Certified.
This item is not returnable. View return policy
Catalog No. NC2101966 Supplier Newcomer Supply Supplier No. 5081W90
The advanced technology in manufacturing the adhesive surface of the TOMO microscope slide produces outstanding adhesion to the tissue section even in the most demanding applications. The surface of the TOMO adhesive microscope slide is hydrophilic producing two very notable features: allows to refloat the tissue in the water bath for repositioning the tissue section and allows the microscope slide to be used on all automated IHC platforms. Glass Type: High Clarity, Low Flouorescence; Size: 75 x 25mm, 1mm thick; Surface Wettability: Hydrophilic; Edge Treatment: 90° Corner; Color: White; Unit: 100 slides/box, 1,000 slides/case. ISO 13485 Certified.