Learn More
Hach Company Tongs, Crucible
Catalog No. 501994395 Shop All Hach Company Products
Encompass
Nickel-plated steel. Length: 230 mm (9"). Wire size: 5 mm.
Catalog No. 50-199-4395 Supplier Hach Company Supplier No. 56900
Nickel-plated steel. Length: 230 mm (9"). Wire size: 5 mm.